Skip to Content
Merck
All Photos(1)

Key Documents

H9378

Sigma-Aldrich

Heptanoic acid

≥97%

Synonym(s):

Enanthic acid, Oenanthic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COOH
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744723
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥97%

expl. lim.

10.1 %

refractive index

n20/D 1.4221 (lit.)

bp

223 °C (lit.)

mp

−10.5 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(O)=O

InChI

1S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

MNWFXJYAOYHMED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carola Buccellati et al.
The Journal of pharmacology and experimental therapeutics, 317(2), 830-837 (2006-01-10)
Thromboxane (TX) A(2), prostacyclin (PGI(2)), and nitric oxide (NO) regulate platelet function and interaction with the vessel wall. Inhibition of TXA(2), implemented synthesis of PGI(2), and supply of exogenous NO may afford therapeutic benefit. 2NTX-99 [4-methoxy-N(1)-(4-trans-nitrooxycyclohexyl)-N(3)-(3-pyridinylmethyl)-1,3-benzenedicarboxamide], a new chemical entity
Dorrain Y Low et al.
Food & function, 6(8), 2464-2474 (2015-07-29)
Fruits (and vegetables) contain cellular structures that are not degraded by human digestive enzymes. Therefore, the structure of the insoluble fraction of swallowed fruits is mostly retained until intestinal microbial fermentation. In vitro fermentation of mango and banana cell structures
L Grislain et al.
European journal of drug metabolism and pharmacokinetics, 15(4), 339-345 (1990-10-01)
After oral administration of amineptine (7-[(10-11)-dihydro-5H-dibenzo(a,d)cycloheptane-5yl] amino heptanoic acid), an original tricyclic antidepressant, seven metabolites were isolated from urine and plasma of rat, dog and man. The metabolic pathways were similar for the three species studied. The two major pathways
Patricia M Jones et al.
Molecular genetics and metabolism, 81(2), 96-99 (2004-01-27)
The treatment for patients with genetic disorders of mitochondrial long-chain fatty acid beta-oxidation is directed toward providing sufficient sources of energy for normal growth and development, and at the same time preventing the adverse effects that precipitate or result from
J M MacLeod et al.
Langmuir : the ACS journal of surfaces and colloids, 31(25), 7016-7024 (2015-01-17)
The behavior of small molecules on a surface depends critically on both molecule-substrate and intermolecular interactions. We present here a detailed comparative investigation of 1,3,5-benzene tricarboxylic acid (trimesic acid, TMA) on two different surfaces: highly oriented pyrolytic graphite (HOPG) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service