Skip to Content
Merck
All Photos(1)

Key Documents

H8004

Sigma-Aldrich

16α-Hydroxyestrone

≥99% (HPLC)

Synonym(s):

1,3,5(10)-Estratriene-3,16α-diol-17-one, 16α-3,16-Dihydroxyestra-1,3,5(10)-trien-17-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O3
CAS Number:
Molecular Weight:
286.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (HPLC)
~95% (TLC)

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)C2=O

InChI

1S/C18H22O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-16,19-20H,2,4,6-7,9H2,1H3/t13-,14-,15+,16+,18-/m0/s1

InChI key

WPOCIZJTELRQMF-UJHHCITNSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Wacker et al.
Journal of women's health (2002), 18(4), 487-491 (2009-04-14)
Hispanic women are at lower risk for incident breast cancer, but the reasons for this lower risk are unknown. Among postmenopausal women, breast cancer risk is inversely associated with circulating levels of 2-hydroxyestrone but directly associated with levels of 16alpha-hydroxyestrone
Susan R Sturgeon et al.
Nutrition and cancer, 62(2), 175-180 (2010-01-26)
Flaxseed is a rich source of dietary lignans. It has been hypothesized that lignans may decrease breast cancer risk through modulation of endogenous hormone levels. The aim of this study was to determine the effect of flaxseed supplementation on urinary
MaryFran R Sowers et al.
Arthritis and rheumatism, 54(8), 2481-2487 (2006-07-28)
To determine if levels of endogenous estrogen or estrogen metabolites are associated with an increased risk of developing knee osteoarthritis (OA) in women. Serum estradiol (E2) and 2 urinary estrogen metabolites (2-hydroxyestrone and 16alpha-hydroxyestrone) with radiographically defined prevalent and incident
Christophe Stavrakakis et al.
Journal of AOAC International, 91(1), 237-246 (2008-04-02)
This paper presents an analytical method applied to the determination of 3 natural steroid hormones, estrone, 17 beta-estradiol, and 16 alpha-hydroxyestrone, and the contraceptive estrogen, 17 alpha-ethynylestradiol, at the sub-ng/L level in water samples [surface water and wastewater treatment plant
MaryFran R Sowers et al.
The Journal of nutrition, 136(6), 1588-1595 (2006-05-17)
Diet and lifestyle factors, body size, and smoking behavior may influence estrogen metabolism, but the nature of these relations may vary according to race/ethnic groups. We evaluated the association of lifestyle factors with estrogen metabolites 2-hydroxyestrone (2-OHE1) and 16alpha-hydroxyestrone (16alpha-OHE1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service