Skip to Content
Merck
All Photos(2)

Documents

H7414

Sigma-Aldrich

Anti-HIP14 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DHHC domain containing 17 (ZDHHC17), Anti-HSPC294, Anti-Huntingtin interacting protein 3 (HIP3), Anti-Huntingtin interacting protein H (HYPH), Anti-Huntingtin-Interacting Protein 14, Anti-KIAA0946, Anti-Zinc finger

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa (mouse)
antigen ~78 kDa (human)

species reactivity

mouse, canine, human, rat, bovine

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

western blot: 0.5-1 μg/mL using extracts of HEK-293T cells expressing human HIP14 and mouse brain extracts (P2 membrane fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Huntingtin interacting protein 14 (HIP14) is a palmitoyl transferase (PAT) neuronal type, localizes to the Golgi, and to vesicles in the cytoplasm. HIP14 mRNA is predominantly expressed in the brain.

Immunogen

synthetic peptide corresponding to amino acids 614-632 located at the C-terminus of human HIP14. This sequence is identical in mouse, rat, dog, and bovine HIP14.

Application

Anti-HIP14 antibody produced in rabbit has been used in quantitative immunoblotting, immunohistochemistry.

Biochem/physiol Actions

The interaction of huntingtin interacting protein 14 (HIP14) with huntingtin protein (htt) is inversely correlated to the polyQ length.. HIP14 along with htt is localized in striatum, medium spiny projection neurons. It is also associated neurons associated with Huntington disease (HD). HIP14 under expression in neurons reduces the clustering of postsynaptic density protein 95 (PSD-95) and glutamic acid decarboxylase 65-kilodalton isoform (GAD65). It has been suggested that HIP14 has a role in palmitoylation and in regulating the intracellular trafficking of multiple neuronal proteins. Decreased interaction between mutant htt and HIP14 could contribute to neuronal dysfunction in HD by perturbing normal intracellular transport pathways in neurons.

Target description

HIP14 (also known as zinc finger, DHHC domain containing protein 17 (ZDHHC17), huntingtin interacting protein H (HYPH), huntingtin interacting protein 3 (HIP3), HSPC294 and KIAA0946) is a novel htt interacting protein.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huntingtin-interacting protein 14 is a type 1 diabetes candidate protein regulating insulin secretion and beta-cell apoptosis
Berchtold LA, et al.
Proceedings of the National Academy of Sciences of the USA, 108(37), E681-E688 (2011)
Neuronal ceroid lipofuscinosis with DNAJC5/CSPalpha mutation has PPT1 pathology and exhibit aberrant protein palmitoylation
Henderson MX, et al.
Acta Neuropathologica, 131(4), 621-637 (2016)
Altered palmitoylation and neuropathological deficits in mice lacking HIP14
Singaraja RR, et al.
Human Molecular Genetics, 20(20), 3899-3909 (2011)
Wild-type HTT modulates the enzymatic activity of the neuronal palmitoyl transferase HIP14
Huang K, et al.
Human Molecular Genetics, 20(17), 3356-3365 (2011)
Protein aggregates in Huntington's disease
Arrasate M and Finkbeiner S
Experimental Neurology, 238(1), 1-11 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service