Skip to Content
Merck
All Photos(1)

Documents

H6643

Sigma-Aldrich

(+)-5(S)-Hydroxy-(6E,8Z,11Z,14Z)-eicosatetraenoic acid

~75 μg/mL in ethanol, ~98%

Synonym(s):

(+)-5-HETE, 5(S)-HETE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O3
CAS Number:
Molecular Weight:
320.47
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~98%

concentration

~75 μg/mL in ethanol

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C/C=C\C=C\[C@@H](O)CCCC(O)=O

Biochem/physiol Actions

A substrate for hydroxyeicosanoid dehydrogenase in polymorphonuclear (PMN) leucocytes, where it is converted to 5-oxo-ETE. Both compounds are reported to increase the intracellular mobilization of calcium in PMN leucocytes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santiago E Farias et al.
The Journal of trauma, 71(5), 1211-1218 (2011-03-24)
Enzymatic and nonenzymatic oxidation of polyunsaturated fatty acids leads to the formation of biologically active products known as lipid mediators. In the brain, lipid mediators play an important role in supporting homeostasis and normal function. Thus, levels of these metabolites
E J Goetzl
Nature, 288(5787), 183-185 (1980-11-13)
The arachidonic acid released from cellular phospholipids of specifically stimulated platelets and leukocytes is oxygenated enzymatically by two major pathways. A complex cycloxygenase converts some of the free arachidonic acid to labile endoperoxides that are transformed to prostaglandins, thromboxanes and
Thorsten J Maier et al.
Biochemical pharmacology, 76(7), 862-872 (2008-08-12)
Celecoxib is a selective cyclooxygenase-2 (COX-2) inhibitor used in the therapy of inflammatory and painful conditions. Various COX-2-independent pharmacological effects, such as a chemo-preventive and tumor-regressive activity have been suggested, but the respective non-COX-2 targets of celecoxib are still a
W S Powell et al.
The Journal of biological chemistry, 267(27), 19233-19241 (1992-09-25)
Human polymorphonuclear leukocytes (PMNL) convert 6-trans isomers of leukotriene B4 (LTB4) to dihydro metabolites (Powell, W.S., and Gravelle, F. (1988) J. Biol. Chem. 263, 2170-2177). In the present study we investigated the mechanism for the initial step in the formation
Stephen R Clark et al.
Blood, 117(6), 2033-2043 (2010-12-24)
5-Lipoxygenase (5-LOX) plays key roles in infection and allergic responses. Herein, four 5-LOX-derived lipids comprising 5-hydroxyeicosatetraenoic acid (HETE) attached to phospholipids (PLs), either phosphatidylethanolamine (PE) or phosphatidylcholine (18:0p/5-HETE-PE, 18:1p/5-HETE-PE, 16:0p/5-HETE-PE, and 16:0a/5-HETE-PC), were identified in primary human neutrophils. They formed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service