H6261
10-Hydroxydecanoic acid
≥95% (GC)
Synonym(s):
NSC 15139
About This Item
Assay
≥95% (GC)
mp
75-77 °C (lit.)
SMILES string
OCCCCCCCCCC(O)=O
InChI
1S/C10H20O3/c11-9-7-5-3-1-2-4-6-8-10(12)13/h11H,1-9H2,(H,12,13)
InChI key
YJCJVMMDTBEITC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Synthesis of copolymers of acrylamide and sodium acryloyloxydecanoate
- Macrolactonization for preparation of Sansalvamide A
- Studies of chain length selectivity for cutinase catalyzed polycondensation reactions
- Ruthenium-catalyzed esterification followed by macrocyclization
- Synthesis of anhydrides and esters
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service