Skip to Content
Merck
All Photos(1)

Key Documents

H3899

Sigma-Aldrich

D-Homocysteinesulfinic acid

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO4S
CAS Number:
Molecular Weight:
167.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

storage temp.

−20°C

SMILES string

NC(CCS(O)=O)C(O)=O

InChI

1S/C4H9NO4S/c5-3(4(6)7)1-2-10(8)9/h3H,1-2,5H2,(H,6,7)(H,8,9)

InChI key

PDNJLMZEGXHSCU-UHFFFAOYSA-N

Biochem/physiol Actions

Potent, fast-acting NMDA glutamate receptor agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Morishita et al.
Journal of neurophysiology, 82(5), 2556-2564 (1999-11-24)
Depolarization-induced suppression of inhibition (DSI) is a process whereby brief approximately 1-s depolarization to the postsynaptic membrane of hippocampal CA1 pyramidal cells results in a transient suppression of GABA(A)ergic synaptic transmission. DSI is triggered by a postsynaptic rise in [Ca(2+)](in)
G A Thompson et al.
Neuropharmacology, 34(8), 857-863 (1995-08-01)
Depolarizations induced by a range of amino acids including some sulphur-containing excitatory transmitter candidates were evoked from motoneurones in the neonatal rat spinal cord under conditions that precluded activation of known ionotropic glutamate receptors. The responses could be partially and
E R Whittemore et al.
European journal of pharmacology, 192(3), 435-438 (1991-01-17)
Quisqualic acid sensitizes hippocampal CA1 neurons to depolarization by L-2-amino-4-phosphonobutanoic acid (L-AP4). This sensitization to L-AP4 is known to be blocked by simultaneous exposure to L-homocysteinesulfinic acid, L-alpha-aminoadipic acid and L-serine-O-sulfate during exposure to quisqualate. We report here that these
Philipp Görtz et al.
Journal of the neurological sciences, 218(1-2), 109-114 (2004-02-05)
Severe hyperhomocysteinemia (50-200 microM) often presents itself with acute neuronal dysfunction including seizures and psychosis. Its moderate form (15-50 microM) is associated with cognitive impairment and dementia. We investigated the neuropharmacological effects of homocysteine and its oxidized forms, homocysteinesulfinic acid
A Becker et al.
International journal of clinical pharmacology and therapeutics, 45(9), 504-515 (2007-10-03)
Interference of methotrexate (MTX) with the metabolism of homocysteine may contribute to MTX neurotoxicity. In this pilot study we measured the concentration of homocysteine and related metabolites in the cerebrospinal fluid (CSF) of patients with primary central nervous system lymphoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service