Skip to Content
Merck
All Photos(1)

Documents

H2255

Sigma-Aldrich

5-Hydroxyindole-3-acetic acid (dicyclohexylammonium) salt

crystalline, ≥98% (TLC)

Synonym(s):

5-HIAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO3 · C12H23N
CAS Number:
Molecular Weight:
372.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

crystalline

color

white

storage temp.

−20°C

SMILES string

C1CCC(CC1)NC2CCCCC2.OC(=O)Cc3c[nH]c4ccc(O)cc34

InChI

1S/C12H23N.C10H9NO3/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;12-7-1-2-9-8(4-7)6(5-11-9)3-10(13)14/h11-13H,1-10H2;1-2,4-5,11-12H,3H2,(H,13,14)

InChI key

NIDSGTAJKGNSEN-UHFFFAOYSA-N

Biochem/physiol Actions

Major serotonin metabolite.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jurjen J Luykx et al.
PloS one, 7(2), e30497-e30497 (2012-02-09)
Animal studies have revealed seasonal patterns in cerebrospinal fluid (CSF) monoamine (MA) turnover. In humans, no study had systematically assessed seasonal patterns in CSF MA turnover in a large set of healthy adults. Standardized amounts of CSF were prospectively collected
ENETS Consensus Recommendations for the Standards of Care in Neuroendocrine Neoplasms: Follow-Up and Documentation.
U Knigge et al.
Neuroendocrinology, 105(3), 310-319 (2017-02-22)
Sofia Raitsin et al.
Brain research, 1666, 1-10 (2017-04-25)
Recent human brain imaging studies implicate dysregulation of monoamine oxidase-A (MAO-A), in particular in the prefrontal cortex (PFC) and anterior cingulate cortex (ACC), in the development of major depressive disorder (MDD). This study investigates the influence of four alterations underlying
Ronghua Fan et al.
Environmental science and pollution research international, 26(21), 22040-22050 (2019-05-31)
Exposure to fine particulate matter (PM2.5) could induce lung impairment aggravation. Moreover, endogenous substances are known to play a significant role in lung impairment. Therefore, the research objectives was to investigate the influence of PM2.5-induced lung impairment on the levels
Xue Tao et al.
International journal of molecular sciences, 21(4) (2020-02-27)
In the pathology-oriented study of depression, inflammation hypothesis has received increasing attention for recent years. To mimic the depressive state caused by inflammation, rodents injected intraperitoneally with lipopolysaccharide (LPS) are usually used to stimulate an immune response. However, the dose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service