Skip to Content
Merck
All Photos(1)

Key Documents

H0790

Sigma-Aldrich

HIV Protease Fluorescent Substrate

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$4,231.00

R$4,231.00


Check Cart for Availability


Select a Size

Change View
1 MG
R$4,231.00

About This Item

Empirical Formula (Hill Notation):
C50H76N14O14
Molecular Weight:
1097.22
UNSPSC Code:
12352204
NACRES:
NA.32

R$4,231.00


Check Cart for Availability

Product Name

HIV Protease Fluorescent Substrate, Ex: 320nm, Em: 420nm

Quality Level

Assay

≥95% (HPLC)

form

lyophilized powder

solubility

acetonitrile: water (1:4): 1 mg/mL

storage temp.

−20°C

Amino Acid Sequence

Abz-Ala-Arg-Val-Nle-Tyr(NO2)-Glu-Ala-Nle-NH2

Biochem/physiol Actions

Substrate for HIV protease (Wildtype and mutants)

Other Notes

Substrate is cleaved between the Nle and Tyr residue, producing a fluorescence that is measured with Ex: 320nm Em: 420nm. This substrate is recognized by both the Wild Type and Drug Resistant Mutants.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service