Skip to Content
Merck
All Photos(1)

Key Documents

GW21146

Sigma-Aldrich

Anti-EGFR antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Epidermal growth factor receptor

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$2,943.00

R$2,943.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
50 μG
R$2,943.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$2,943.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-21146

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)

Immunogen

Immunogen Sequence: GI # 29725609, sequence 1091-1210
Recombinant epidermal growth factor receptor isoform a

Application

Anti-EGFR antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

The epidermal growth factor receptor (EGFR) is a transmembrane glycoprotein and belongs to erbB family of tyrosine kinase receptors. It binds to its cognate ligands that leads to autophosphorylation of receptor tyrosine kinase and subsequent activation of signal transduction pathways that are involved in regulating cellular proliferation, differentiation and survival. EGFR is overexpressed in tumor cell lines and is associated with poor prognosis leading to decreased survival. Its activation also plays a role in resistance to chemotherapy and radiation treatment in tumor cells. EGFR tyrosine kinase inhibitors (TKIs) may act as a new treatment option for advanced lung adenocarcinoma. Its ligands function in the Drosophila CNS and maintains glial survival. Drosophila EGF receptor plays a critical role during the development of Drosophila retina.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roy S Herbst
International journal of radiation oncology, biology, physics, 59(2 Suppl), 21-26 (2004-05-15)
The epidermal growth factor receptor (EGFR) is a transmembrane glycoprotein that constitutes one of four members of the erbB family of tyrosine kinase receptors. Binding of EGFR to its cognate ligands leads to autophosphorylation of receptor tyrosine kinase and subsequent
Shiue-Wei Lai et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 19(2), 459-465 (2014-06-27)
Epidermal growth factor receptor (EGFR) tyrosine kinase inhibitors (TKIs) represent a new treatment option for patients with advanced lung adenocarcinoma. In this article we assessed the treatment response and tried to identify prognostic factors which may provide some information different
Alicia Hidalgo
Trends in neurosciences, 25(7), 365-370 (2002-06-25)
The non-autonomous control of cell survival has long been thought to be a mechanism of adjusting cell populations in the vertebrate nervous system, enabling connectivity and myelination to produce a functional brain. Despite cellular evidence that analogous mechanisms occur in
S A Spencer et al.
Development (Cambridge, England), 125(23), 4777-4790 (1998-11-10)
Developing epithelia use a variety of patterning mechanisms to place individual cells into their correct positions. However, the means by which pattern elements are established are poorly understood. Here, we report evidence that regulation of Drosophila EGF receptor (DER) activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service