Key Documents
GN782078
Greiner CELLSTAR® microplate
1536 well, HiBase, TC treated, black
Synonym(s):
1536 well microplate, 1536 well plate, cell culture plate, treated microplate
About This Item
UNSPSC Code:
41122107
material:
black
size:
1536 wells
Pricing and availability is not currently available.
material
black
packaging
pack of 60 ea
manufacturer/tradename
Greiner 782078
availability
available only in China
size
1536 wells
suitability
suitable for (cell culture)
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)