Key Documents
GN781090
Greiner CELLSTAR® microplate
384 well, TC treated, black μClear®
Synonym(s):
384 well microplate, 384 well plate, cell culture plate, treated microplate
About This Item
UNSPSC Code:
41122107
material:
black
size:
384 wells
Pricing and availability is not currently available.
material
black
packaging
pack of 120 ea
manufacturer/tradename
Greiner 781090
availability
available only in China
size
384 wells
suitability
suitable for (cell culture)
General description
  • Greiner CELLSTAR® 384 Well Cell Culture Plates enable improved cell adhesion through physical surface treatment.
  • Features a lid that facilitates gas exchange with the lowest possible evaporation and significantly reduces the risk of cross-contamination.
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic and non-cytotoxic.
  • μClear® bottom is a transparent thin-film bottom ideal for cell culture and microscopic applications using fluorescence or luminescence detection methods.
  • A rounded square well profile facilitates flexible working volume and prevents bubbling.
  • Suitable for all common instruments and compatible with automated systems.
  • Alphanumeric well coding.
Legal Information
μClear is a registered trademark of Greiner Bio-One GmbH
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)