Skip to Content
Merck
All Photos(1)

Key Documents

GN662102

Greiner CELLSTAR® multiwell culture plates

24 well, suspension

Synonym(s):

culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
material:
polystyrene

material

polystyrene

sterility

sterile

packaging

pack of 100 ea

manufacturer/tradename

Greiner 662102

availability

available only in China

Looking for similar products? Visit Product Comparison Guide

Application

Greiner CELLSTAR® multiwell culture plates are suitable for suspension culture cells and hybridoma cells.

Features and Benefits

  • Detectable DNase-, RNase-, human DNA-free
  • Non-pyrogenic and non-cytotoxic
  • The lid enables gas exchange with minimal evaporation
  • Single-position lids prevent cross-contamination
  • Well coding with alphanumeric values
  • Packaging is easy to open
  • Compatible with automated systems
  • Suitable for automation and all common instruments
  • High clarity and low autofluorescence
  • Hydrophobic surface

Legal Information

CELLSTAR is a registered trademark of Greiner Bio-One GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service