Skip to Content
Merck
All Photos(1)

Key Documents

GE90100484

Streptavidin Sepharose High Performance

Cytiva, 90100484, pkg of 25 mL

Synonym(s):

Streptavidin resin

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
R$26,147.00

R$26,147.00


Check Cart for Availability


Select a Size

Change View
25 ML
R$26,147.00

About This Item

UNSPSC Code:
41106500
NACRES:
NB.21

R$26,147.00


Check Cart for Availability

packaging

pkg of 25 mL

manufacturer/tradename

90100484
Cytiva

Looking for similar products? Visit Product Comparison Guide

General description

Streptavidin Sepharose High Performance is an affinity chromatography resin designed for fast and reliable, high resolution purification of biotinylated biomolecules. Purified Streptavidin isolated from Streptomyces avidinii is immobilised on Sepharose High Performance. The base matrix is a rigid, highly cross-linked beaded agarose with high chemical stability. The immobilised streptavidin binds biotin and biotinylated substances through affinity chromatography.

Features and Benefits

  • Streptavidin immobilized to Sepharose High Performance for purification of biotinylated compounds
  • Extremely useful for exploiting either the strong interactions of biotin and streptavidin or the somewhat weaker interactions of 2-iminobiotin and streptavidin.
  • 34 µm bead size for improved resolution

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service