Skip to Content
Merck
All Photos(1)

Documents

GE29321906

Superdex® 200 Increase HiScale®

column thread × L (26/40) (40 cm), Cytiva 29321906

Synonym(s):

Superdex 200 Column, Superdex 200 Increase Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
NB.21

feature

Sample volume up to 8.5 mL

manufacturer/tradename

Cytiva 29321906

parameter

2 Mpa max. pressure

bed volume × D × L

(212 mL) (26 mm) (400 mm)

column thread × L

(26/40) (40 cm)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

HiScale columns, packed with new-generation SEC resins, provide the highest resolution among preparative SEC columns from Cytiva.

If you like our new-generation Superdex Increase and Superose Increase size exclusion chromatography (SEC) columns, but need to purify larger volumes of up to 8.5 mL, we can pack the resin for you in HiScale columns.

HiScale columns are packed on demand for you with Superdex 200 Increase, Superdex 75 Increase, and Superose 6 Increase SEC (gel filtration) resins. The columns are available in two sizes: 16 × 400 mm and 26 × 400 mm. These two column sizes enable purification of sample volumes up to 3.2 and 8.5 mL, respectively.

Features and Benefits

  • High-resolution SEC columns—small particle size and narrow particle size distribution of the packed resin ensures highly pure protein.
  • HiScale columns packed with SEC resins on demand, please request a quote.

Legal Information

HiScale is a registered trademark of Cytiva
Superdex is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

96.8 °F

Flash Point(C)

36 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service