Key Documents
GE29053586
Tricorn Filter Kit
Tricorn 5 Columns
About This Item
UNSPSC Code:
41104925
NACRES:
NB.43
Pricing and availability is not currently available.
description
Tricorn glass columns may be used with aqueous buffer solutions and detergents at pH conditions between 1 and 14, and with nearly all organic solutions commonly used in chromatography.
feature
Ethylene propylene diene (EPDM) Polyethylene (PE)
Kit Contents: 5 x Top filter 5 x Bottom filter 5 x O-ring 7 x 1 mm (EPDM)
Tricorn 5 Columns
Wetted Material: Ethylene propylene diene (EPDM) Polyethylene (PE)
manufacturer/tradename
Cytiva 29053586
parameter
4 °C min. temp.
60 °C max. temp.
General description
Filter kits for Tricorn 5 and 10 columns are available in three porosities. Choose the standard 7 µm porosity for most applications. Use the medium 18 µm porosity for the purification of polyclonal IgG antibodies or mAbs in combination with resins such as MabSelect SuRe pcc or MabSelect PrismA. Use the coarse filter (> 25 µm) to reduce the risk of clogging your capture resin when working with large volumes or repeated loadings of clarified samples. All filter kits include top and bottom filters and ethylene propylene diene monomer (EPDM) O-rings, five of each.
Tricorn columns incorporate modern design and high performance to increase the user friendliness and reliability of the chromatography column. Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures.
Features and Benefits
  • High performance glass columns designed for reliable high resolution, reproducibility and durability
  • New longer adaptor for increased adjustment of bed height
  • Modern, integrated design – simple to assemble, pack, operate, and maintain
  • Broad range of column dimensions – suitable for many different purification needs
  • Safety first – glass tubes coated with plastic film to protect laboratory personnel if the column is dropped or operated outside its specifications
  • Locking ring – prevents accidental compression of media bed
  • Separate packing connector – allows simple column packing for a broad range of media
Choose from one of the most recent versions:
Certificates of Analysis (COA)