GE27-4110-01
Poly (A)
Cytiva 27-4110-01, pack of 100 mg
Synonym(s):
Polyadenylated RNA, Polyadenylic acid
About This Item
Recommended Products
packaging
pack of 100 mg
manufacturer/tradename
Cytiva 27-4110-01
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Biochem/physiol Actions
Storage and Stability
Analysis Note
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service