GE17545908
HiTrap® Capto™ Butyl
pkg of 5 × 1 mL, Cytiva, 17-5459-08, 75 μm avg. part. size
Select a Size
R$3,007.00
Select a Size
About This Item
R$3,007.00
Recommended Products
packaging
pkg of 5 × 1 mL
manufacturer/tradename
17-5459-08
Cytiva
avg. part. size
75 μm
General description
- Aliphatic ligand coupled with a flexible spacer for laboratory and process applications requiring a HIC resin with low to resin hydrophobicity
- Highly rigid matrix enables high flow rates, long bed heights and faster throughput
- Improves productivity and process economy in downstream operations
- Narrow specification range ensures robust performance in sensitive processes
- Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service