Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1355-01

HiTrap® Phenyl Fast Flow (HS)

Cytiva 17-1355-01, pack of 5 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

phenyl

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-1355-01

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning

2-14

working range

3-13

suitability

suitable for bioprocess medium

General description

HiTrap® Phenyl FF (high sub) are prepacked Phenyl Sepharose 6 Fast FLow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elutions conditions.

Features and Benefits

  • Provide convenient and fast small-scale Protein purifications.
  • Phenyl Sepharose 6 Fast FLow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring Low to medium hydrophobicity. Two levels of substitution on Phenyl Sepahrose 6 Fast FLow help find optimal selectivity and binding capacity.
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA® design.
  • High flow rates and high capacities.
  • Excellent and easy scale-up achieved by connecting several HiTrap® columns in series or using HiPrep columns.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiPrep is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

This page shows how to remove serine proteases, e.g. thrombin and trypsin, and zymogens from recombinant products using HiTrap Benzamidine FF and Benzamidine Sepharose 4 Fast Flow from Cytiva.

Column, media, and sample preparation for Hydrophobic Interaction Chromatography (HIC) detailed with Cytiva media.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

This page shows how to purify ATPases, adenylate cyclases, protein kinases, phosphodiesterases and neurotransmitters by affinity chromatography using Calmodulin Sepharose 4B from Cytiva.

This page shows how to condition membrane proteins for further analysis with products from Cytiva.

This page discusses various aspects of sample preparation for chromatographic purification.

This page shows how to perform column packing and preparation for affinity chromatography when using Tricorn™ or XK columns available from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service