Skip to Content
Merck
All Photos(2)

Key Documents

GE17-0920-05

HiTrap® IMAC High Performance

Cytiva 17-0920-05, pack of 5 × 5 mL

Synonym(s):

Metal affinity chromatography

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56
Pricing and availability is not currently available.

packaging

pack of 5 × 5 mL

manufacturer/tradename

Cytiva 17-0920-05

parameter

42 psi (H2O at room temperature.)

bed size

16 mm × 25 mm

bed volume

5 mL

column I.D.

16 mm

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-14(Medium uncharged with metal ions.)

working range

3-12

capacity

≥40 mg binding capacity (histidine-tagged protein/ml medium when changed with Ni2+)(Protein- and metal-ion dependent.)

General description

HiTrap® IMAC HP is prepacked with IMAC Sepharose High Performance. The medium is charged with the metalion of your choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent high-resolution purification of polyhistidine tagged proteins.

Application

IMAC Sepharose HP is supplied free of metal ions. It is charged by the user with the transition metal ion of choice (e.g. Cu2+, Zn2+, Ni2+, or Co2+); these metal ions will bind to the covalently immobilized chelating ligand on the Sepharose.

Features and Benefits

  • For optimizing purification of histidine-tagged Proteins when Ni2+ is not the best choice of metal ion.
  • Conveniently charge with your metal of choice.
  • Small bead size for high-performance purification.
  • High binding capacity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

IMAC Sepharose products, Ni Sepharose products and Fe Sepharose products
These products are sold under a license from Sigma-Aldrich under patent number EP 1276716 (Metal chelating compositions) and equivalent patents and patent applications in other countries.
HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

How to optimize purification of histidine-tagged proteins using Cytiva products.

This page shows troubleshooting instructions for affinity chromatography of tagged proteins using Cytiva products.

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page describes manual and automated purification of histidine-tagged recombinant proteins using Cytiva products

Protocols

This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service