Skip to Content
Merck
All Photos(1)

Documents

GE17-0310-50

Ficoll® PM 70

Cytiva 17-0310-50, pack of 500 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

form

solid
spray-dried

packaging

pack of 500 g

manufacturer/tradename

Cytiva 17-0310-50

color

White

solubility

soluble

General description

High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
It is delivered as a spray-dried powder and is therefore readily soluble in aqueous media when it is added slowly with concomitant stirring. Any lumps formed during this procedure will dissolve fairly quickly. Concentrations up to 50% (w/v) can easily be attained

Features and Benefits

  • Displays minimal toxicity
  • Has favorable viscosity and osmotic properties

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Ficoll is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service