GE10600032
Amersham™ Protran® Western blotting membranes, nitrocellulose
pore size 0.2 μm, sheet W × L 300 mm × 600 mm, pkg of 5 ea
About This Item
Recommended Products
packaging
pkg of 5 ea
manufacturer/tradename
Cytiva 10600032
sheet W × L
300 mm × 600 mm
pore size
0.2 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Western blotting
- Southern blotting
- Northern blotting
Features and Benefits
- Low background and excellent signal-to-noise ratios, without the need for stringent washing conditions systems.
- Compatible with a variety of Western blot labeling and detection systems
- Risk of “blow-through” reduced by high binding capacity of small proteins (< Mr 20 000)
- Optimized for chemiluminescent and fluorescent detection
- Long shelf life of bound proteins to the transfer membrane
- Available in popular precut sizes for Western blot transfer applications
Storage and Stability
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service