GE10600017
Amersham™ Protran® Supported Western blotting membranes, nitrocellulose
pore size 0.2 μm, roll W × L 200 mm × 4 m, pkg of 1 ea
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 10600017
roll W × L
200 mm × 4 m
pore size
0.2 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Western blotting
- Southern blotting
- Northern blotting
Features and Benefits
- High binding capacity.
- Versatile, robust and excellent signal-to-noise ratios.
- Optimal for applications when stripping and reprobing is required.
- Suitable for proteins of a wide range of molecular weights.
Storage and Stability
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service