GBL475023
Grace Bio-Labs CoverWell™ imaging chambers
diam. × thickness 9 mm × 1.6 mm
Synonym(s):
leak proof chamber, leak proof enclosure, microscope stick on chamber, reusable press to seal chamber
Select a Size
R$1,191.00
Select a Size
About This Item
R$1,191.00
Recommended Products
sterility
non-sterile
feature
wells 1
packaging
pack of 25 ea
manufacturer/tradename
Grace Bio-Labs 475023
diam. × thickness
9 mm × 1.6 mm
Looking for similar products? Visit Product Comparison Guide
General description
The silicone chambers form removable, water-tight chambers to support thick and free-floating specimens for imaging. Flexible coverslip material allows these chambers to be removed from the thinnest coverglass without breaking the glass or damaging the specimen. The thin (0.18mm / 0.007”) polycarbonate cover displays excellent transparency. Imaging chambers are available with or without adhesive. Adhesive CoverWells are designed to form peel-and-stick leak-proof enclosures for the permanent mounting of thick specimens.
Application
- Confocal microscopy
- Imaging
- Tissue and Cell staining
- High Resolution Microscopy
- Live-cell imaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service