Key Documents
GBL246824
Grace Bio-Labs ProPlate® microarray system tray set
well size 24, Square, With Stainless Steel Spring Clips
Synonym(s):
incubation chambers, microarray hybridization chambers, slide chambers
Select a Size
1 EA
R$2,482.00
R$2,482.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$2,482.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
R$2,482.00
Check Cart for Availability
material
black silicone (Acrylic tray)
feature
Includes Tray and Cover, 10 Seal Strips, and 1 Applicator
Square, With Stainless Steel Spring Clips
packaging
pack of 1 ea
manufacturer/tradename
Grace Bio-Labs 246824
well size
24
General description
ProPlate re-usable, multi-well chambers form removable, leak-proof wells on virtually any slide surface without the use of adhesive.
Designs feature ANSI-SLAS compliant well spacing to facilitate multichannel pipetting.
Multiple slides insert easily into frames having a standard microtiter plate footprint for automated processing.
Legal Information
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)