Skip to Content
Merck
All Photos(2)

Documents

GBL104212

Grace Bio-Labs SecureSlip silicone supported coverglass

diam. 12 mm

Synonym(s):

glass coverslips, silicone backed coverglass

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

material

borosilicate glass cover
colorless
silicon chamber

sterility

(sterile)
sterile

feature

wells 15 (in each sealed pack)

manufacturer/tradename

Grace Bio-Labs 104212

packaging

pkg of (Shipped as 5 sealed packs each containing 15 chambers)

diam.

12 mm

external L × W

130 mm × 72 mm

well area

1.1 cm2

wells

15

Looking for similar products? Visit Product Comparison Guide

General description

The SecureSlip coverglass is affixed to a thin microscopically transparent silicone base which secures to culture vessels by an electrostatic charge, not an adhesive, to prevent movement during cultivation. The silicone also acts as a hydrophobic barrier to isolate liquid reagents on glass. Individual coverglass with silicone backing can be adhered to a petri dish or plate.

These can be easily removed one at a time from the dish during culture. Once removed, coverglass can be affixed to a microscope slide by means of the tacky silicone backing for routine processing, or placed cell side down onto a drop of reagent using the silicone backing as a cover to prevent evaporation.

  • Silicone backing peels off easily for conventional mounting on microscope slides
  • During culture, cells may be imaged without interference from the optical grade silicone
  • Eliminates coverglass overlapping during culture
  • Easily identify "cell side" of coverglass
  • Recommended use temperature is -62 °C to 218 °C

Application

Cell Culture
Immunofluorescence
Microscopy

Legal Information

SecureSlip is a trademark of Grace Bio-Labs, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service