Skip to Content
Merck
All Photos(1)

Key Documents

GBL103201

Grace Bio-Labs Culturewell silicone well dividers

wells, 2, L × W × D 18 mm × 18 mm × 1.6 mm, sterile, pack of 4 ea

Synonym(s):

silicone 2 well divider, slide well divider

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
R$976.00

R$976.00


Check Cart for Availability


Select a Size

Change View
4 EA
R$976.00

About This Item

UNSPSC Code:
41161502
NACRES:
NB.17

R$976.00


Check Cart for Availability

sterility

sterile

feature

wells 2

packaging

pack of 4 ea

manufacturer/tradename

Grace Bio-Labs 103201

L × W × D

18 mm × 18 mm × 1.6 mm

General description

Reusable silicone well dividers designed to affix to the surface of Culturewell coverslip gaskets, isolating desired reagents inserted into the openings. Culturewell silicone dividers are designed to affix to the surface of Culturewell coverslip gaskets. Reagents may be pipetted into the openings of well dividers as desired. Silicone well dividers may be re-used by rinsing thorough with distilled water after each use and re-sterilization. Dividers may be sterilized by autoclaving, or alcohol rinse. Unused sterile dividers may be stored by placing them in sterile, covered culture dish until ready to use.

Legal Information

Culturewell is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service