Skip to Content
Merck
All Photos(1)

Key Documents

G9380

Sigma-Aldrich

α-D-Glucose 1-phosphate disodium salt hydrate

98-99%

Synonym(s):

α-D-Glucopyranose 1-phosphate, Cori ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11O9PNa2 · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

98-99%

form

powder

technique(s)

thin layer chromatography (TLC): suitable

impurities

α-D-glucose 1,6-diphosphate, glucose and starch, essentially free

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H]O[H].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H13O9P.2Na.H2O/c7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13;;;/h2-10H,1H2,(H2,11,12,13);;;1H2/q;2*+1;/p-2/t2-,3-,4+,5-,6-;;;/m1.../s1

InChI key

YWAUQXHOCBBYLP-FBNUBEQJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

α-D-Glucose 1-phosphate is converted into D-glucose-6-phosphate by the enzyme phosphoglucomutase.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanley A Blumenthal
Perspectives in biology and medicine, 55(2), 236-249 (2012-05-31)
In 1945, Earl Sutherland (1915-1974) [corrected] and associates began studies of the mechanism of hormone-induced glycogen breakdown in the liver. In 1956, their efforts culminated in the identification of cyclic AMP, an ancient molecule generated in many cell types in
Ting Yang et al.
The Biochemical journal, 429(3), 533-543 (2010-05-21)
The diverse types of glycoconjugates synthesized by trypanosomatid parasites are unique compared with the host cells. These glycans are required for the parasite survival, invasion or evasion of the host immune system. Synthesis of those glycoconjugates requires a constant supply
Jef Van der Borght et al.
Biotechnology journal, 5(9), 986-993 (2010-08-28)
β-D-Glucose-1-phosphate (βGlc1P) is an efficient glucosyl donor for both enzymatic and chemical glycosylation reactions but is currently very costly and not available in large amounts. This article provides an efficient production method of βGlc1P from trehalose and phosphate using the
Bijay Singh et al.
Protein engineering, design & selection : PEDS, 25(4), 179-187 (2012-02-16)
Two similar genes, dnmL and rmbA in Streptomyces peucetius, which encode for glucose-1-phosphate (G-1-P) thymidylyltransferases were expressed in Escherichia coli under similar conditions. While RmbA was expressed in soluble form, DnmL was found as insoluble aggregates in inclusion bodies. The
Joerg Fettke et al.
Plant physiology, 155(4), 1723-1734 (2010-12-01)
Almost all glucosyl transfer reactions rely on glucose-1-phosphate (Glc-1-P) that either immediately acts as glucosyl donor or as substrate for the synthesis of the more widely used Glc dinucleotides, ADPglucose or UDPglucose. In this communication, we have analyzed two Glc-1-P-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service