Skip to Content
Merck
All Photos(1)

Key Documents

G7798

Sigma-Aldrich

GYKI-53655 hydrate

>99.0% (HPLC)

Synonym(s):

1-(4-aminophenyl)-3-methylcarbamyl-4-methyl-3,4-dihydro-7,8-methylenedioxy-5H-2,3-benzodiazepine hydrochloride, 5-(4-Aminophenyl)-8,9-dihydro-N,8-dimethyl-7H-1,3-dioxolo[4,5-h][2,3]benzodiazepine-7-carboxamide hydrochloride, LY300168 hydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,779.00
25 MG
R$6,578.00

R$1,779.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$1,779.00
25 MG
R$6,578.00

About This Item

Empirical Formula (Hill Notation):
C19H20N4O3 · HCl · xH2O
CAS Number:
Molecular Weight:
388.85 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$1,779.00


Check Cart for Availability

Request a Bulk Order

Assay

>99.0% (HPLC)

form

powder

color

yellow to orange

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

[Cl-].N1(N=C(c3c(cc4c(c3)OCO4)CC1C)c2ccc(cc2)N)C(=O)NC.[H+]

InChI

1S/C19H20N4O3.ClH/c1-11-7-13-8-16-17(26-10-25-16)9-15(13)18(22-23(11)19(24)21-2)12-3-5-14(20)6-4-12;/h3-6,8-9,11H,7,10,20H2,1-2H3,(H,21,24);1H

InChI key

ASLCSBBDVWPSQT-UHFFFAOYSA-N

Application

GYKI-53655 hydrate has been used as a calcium-permeable α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPA) receptor antagonist in single-particle tracking experiments.[1]

Biochem/physiol Actions

GYKI-53655 also inhibits the kainate GluK3 homomeric and GluK2/3 heteromeric receptors.[2] It exhibits anti-ischemic, anti-convulsant, and acute neuroprotective activity.[3]
GYKI-53655 is an AMPA antagonist and noncompetitive negative allosteric modulator.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Szabados et al.
Brain research bulletin, 55(3), 387-391 (2001-08-08)
GYKI 52466 [1-(4-aminophenyl)-4-methyl-7,8-methylenedioxy-5H-2,3-benzodiazepine], a non-competitive AMPA [alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionate] and kainate receptor antagonist and its two analogues, GYKI 53405 [1-(4-aminophenyl)-3-acetyl-4-methyl-3,4-dihydro-7,8-methylenedioxy-5H-2,3-benzodiazepine] and GYKI 53655 [1-(4-aminophenyl)-3-methylcarbamyl-4-methyl-3,4-dihydro-7,8-methylenedioxy-5H-2,3-benzodiazepine] were investigated in two seizure models and in MgCl2 induced global cerebral ischaemia, as an acute neuroprotective model.
David Perrais et al.
Neuropharmacology, 56(1), 131-140 (2008-09-02)
A number of kainate receptor antagonists have shown selectivity for receptors containing the GluK1 subunit. Here, we analyze the effects of these GluK1 antagonists on currents mediated by recombinant homomeric GluK3 and heteromeric GluK2/3 receptors expressed in HEK 293 cells
Alice Polenghi et al.
Cell reports, 31(10), 107735-107735 (2020-06-11)
Kainate receptors (KARs) mediate postsynaptic currents with a key impact on neuronal excitability. However, the molecular determinants controlling KAR postsynaptic localization and stabilization are poorly understood. Here, we exploit optogenetic and single-particle tracking approaches to study the role of KAR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service