Skip to Content
Merck
All Photos(1)

Key Documents

G7673

Sigma-Aldrich

α-Galactosidase I, Alkaline from Cucumis melo

Synonym(s):

α-D-Galactoside Galactohydrolase, Melibiase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

lyophilized solid

specific activity

≥20 units/mg protein

mol wt

apparent mol wt ~84 kDa by SDS-PAGE

shipped in

wet ice

storage temp.

−20°C

General description

Alkaline α-Galactosidase I is a glycoside hydrolase, separated from the extracts of melon fruits. The activity of this enzyme is found to increase during the early stages of ovary development and fruit set.

Application

Alkaline α-Galactosidase I was used to assay enzyme activity with 2 mmp-nitrophenyl-α-d-galactoside as substrate at pH 6.5 to compare with the enzyme activity of α-Gal A isolated and purified from Sf-9 insect cells infected with a recombinant baculovirus encoding normal α-Gal A gene.

Biochem/physiol Actions

α-Galactosidase form I from melon may play a significant role in photoassimilate partitioning in sink tissue. The form I enzyme has a preferred activity with raffinose and significant activity with stachyose. A unique feature of this enzyme is that it exhibits weak product inhibition by galactose.
This enzyme shows a preference for raffinose and also activity with stachyose. Unlike other a-Galctosidases, this enyzme also shows weak inhibition by galactose.

Physical properties

This enzyme exhibits maximal activity between 30-37°C. Activity declines above 40°C.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl α-D-galactoside to p-nitrophenol and D-galactose per minute at pH 7.8 at 30 °C.

Physical form

The product is supplied as a lyophilized powder containing Tris-HCl buffer salts, DTT, EDTA, and NaCl.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Asano et al.
European journal of biochemistry, 267(13), 4179-4186 (2000-06-24)
Fabry disease is a lysosomal storage disorder caused by deficient lysosomal alpha-galactosidase A (alpha-Gal A) activity. Deficiency of the enzyme activity results in progressive deposition of neutral glycosphingolipids with terminal alpha-galactosyl residue in vascular endothelial cells. We recently proposed a
Gao et al.
Plant physiology, 119(3), 979-988 (1999-03-09)
The cucurbits translocate the galactosyl-sucrose oligosaccharides raffinose and stachyose, therefore, alpha-galactosidase (alpha-D-galactoside galactohydrolase, EC 3.2.1.22) is expected to function as the initial enzyme of photoassimilate catabolism. However, the previously described alkaline alpha-galactosidase is specific for the tetrasaccharide stachyose, leaving raffinose
Nir Carmi et al.
The Plant journal : for cell and molecular biology, 33(1), 97-106 (2003-08-29)
Raffinose and stachyose are ubiquitous galactosyl-sucrose oligosaccharides in the plant kingdom which play major roles, second only to sucrose, in photoassimilate translocation and seed carbohydrate storage. These sugars are initially metabolised by alpha-galactosidases (alpha-gal). We report the cloning and functional
Dominique P Germain et al.
Orphanet journal of rare diseases, 7, 91-91 (2012-11-28)
Fabry disease (FD) is a genetic disorder resulting from deficiency of the lysosomal enzyme α-galactosidase A (α-Gal A), which leads to globotriaosylceramide (GL-3) accumulation in multiple tissues. We report on the safety and pharmacodynamics of migalastat hydrochloride, an investigational pharmacological
Costanza Simoncini et al.
The neurologist, 18(6), 413-414 (2012-11-02)
Fabry disease (FD) is a rare, X-linked lysosomal storage disorder with multiorgan involvement. FD is caused by a partial or total deficit of α-galactosidase A enzyme, which is responsible for the accumulation of glycosphingolipids in a variety of cell types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service