G5754
Glyoxal trimer dihydrate
≥97%
Synonym(s):
Ethanedial trimer dihydrate, Hexahydro[1,4]dioxino[2,3-b]-1,4-dioxin-2,3,6,7-tetrol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(HC(O)C(O)H)3 · 2H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
3542160
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
≥97%
SMILES string
[H]O[H].[H]O[H].OC1OC2OC(O)C(O)OC2OC1O
InChI
1S/C6H10O8/c7-1-2(8)12-6-5(11-1)13-3(9)4(10)14-6/h1-10H
InChI key
BDSPTFQIOAEIII-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reactant involved in the synthesis of:
- Aza-analogs of 1,4-naphthoquinones as effectors of plasmodial thioredoxin and glutathione reductase
- Substituted side-bridged cyclam derivatives
- Furandicarboxylic acid di-methyl ester derivatives
Other Notes
This form of glyoxal is composed of 3 moles of glyoxal and 2 moles of water in a relatively stable configuration.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>239.0 °F
Flash Point(C)
> 115 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service