Skip to Content
Merck
All Photos(1)

Key Documents

G3252

Sigma-Aldrich

Gly-Ser

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O4
CAS Number:
Molecular Weight:
162.14
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

[H][C@](CO)(NC(=O)CN)C(O)=O

InChI

1S/C5H10N2O4/c6-1-4(9)7-3(2-8)5(10)11/h3,8H,1-2,6H2,(H,7,9)(H,10,11)/t3-/m0/s1

InChI key

BCCRXDTUTZHDEU-VKHMYHEASA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabeth Häusler et al.
Biochimica et biophysica acta. Biomembranes, 1862(4), 183190-183190 (2020-01-15)
A membrane protein's oligomeric state modulates its functionality in various cellular processes. Since membrane proteins have to be solubilized in an appropriate membrane mimetic, the use of classical biophysical methods to analyze protein oligomers is challenging. We here present a
Di Wu et al.
Scientific reports, 9(1), 6172-6172 (2019-04-18)
Many cellular functions, including signaling and regulation, are carried out by intrinsically disordered proteins (IDPs) binding to their targets. Experimental and computational studies have now significantly advanced our understanding of these binding processes. In particular, IDPs that become structured upon
Michelle L Parker et al.
PloS one, 10(5), e0126206-e0126206 (2015-05-09)
Apicomplexan parasites are the causative agents of globally prevalent diseases including malaria and toxoplasmosis. These obligate intracellular pathogens have evolved a sophisticated host cell invasion strategy that relies on a parasite-host cell junction anchored by interactions between apical membrane antigens
Hikaru Nakazawa et al.
Protein and peptide letters, 27(6), 557-566 (2019-11-16)
By recent advances in phage-display approaches, many oligopeptides exhibiting binding affinities for metal oxides have been identified. Indium tin oxide is one of the most widely used conductive oxides, because it has a large band gap of 3.7-4.0 eV. In
Ling Zhang et al.
Food & function, 8(4), 1698-1707 (2017-03-08)
Applying antioxidants to attenuate skin photoaging has received great attention. In this study, antioxidant collagen peptides (ACPs) with different activities were prepared using different proteases, called high (HCP), medium (MCP) and low antioxidant collagen peptides (LCP). The effects of ACPs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service