Skip to Content
Merck
All Photos(1)

Key Documents

G1765

Sigma-Aldrich

Glycogen from oyster

Type XI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

biological source

animal (oyster)

type

Type XI

form

powder

application(s)

cell analysis

storage temp.

2-8°C

InChI

1S/C24H42O21/c25-1-5-9(28)11(30)16(35)22(41-5)39-4-8-20(45-23-17(36)12(31)10(29)6(2-26)42-23)14(33)18(37)24(43-8)44-19-7(3-27)40-21(38)15(34)13(19)32/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15-,16-,17-,18-,19-,20-,21+,22+,23-,24-/m1/s1

InChI key

BYSGBSNPRWKUQH-UJDJLXLFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycogen is a branched polymer of glucose synthesized by animal cells for energy storage and release. It is constructed of predominantly α1→4 glycosidic bonds with branches created through α1→6 glycosidic bonds.

Application

Oyster glycogen has been used to induce inflammation and to promote extravasation of granulocytes and neutrophils in animal models after intraperitoneal injection. Glycogen also acts as a vascular permeability probe.

Other Notes

High molecular weight

Preparation Note

Prepared by a modification of Bueding, E., and Orrell, S.A., Jr., J. Biol. Chem., 236, 2854 (1961).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Dus et al.
Nature neuroscience, 16(5), 526-528 (2013-04-02)
Animals can determine the nutritional value of sugar without the influence of taste. We examined a Drosophila mutant that is insensitive to the nutritional value of sugars, responding only to the concentration (that is, sweetness). The affected gene encodes a
Marta Julia Faro et al.
Experimental parasitology, 134(2), 228-234 (2013-04-02)
Parasitic castration in the snail-trematode relationship can be understood as any change in the reproductive function of the snail that is due to interference by the developing larvae inside the snail that leads to the reduction or complete disruption of
Jith K Thomas et al.
Aquatic toxicology (Amsterdam, Netherlands), 130-131, 112-122 (2013-02-13)
In a previous study we reported impaired swimming performance and greater stored energy in adult zebrafish (Danio rerio) after chronic dietary exposure to selenomethionine (SeMet). The goal of the present study was to further investigate effects of chronic exposure to
Joachim Nielsen et al.
Applied physiology, nutrition, and metabolism = Physiologie appliquee, nutrition et metabolisme, 38(2), 91-99 (2013-02-27)
Glucose is stored in skeletal muscle fibers as glycogen, a branched-chain polymer observed in electron microscopy images as roughly spherical particles (known as β-particles of 10-45 nm in diameter), which are distributed in distinct localizations within the myofibers and are
Juan M Pascual et al.
JAMA neurology, 70(6), 756-763 (2013-04-24)
The physiological relevance of acid maltase (acid α-glucosidase, an enzyme that degrades lysosomal glycogen) is well recognized in liver and muscle. In late (adult)-onset acid maltase deficiency (glycogen storage disease type II [GSD II]), glycogen accumulates inside muscular lysosomes in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service