Skip to Content
Merck
All Photos(1)

Key Documents

G1651

Sigma-Aldrich

[des-His1, Glu9]-Glucagon amide

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C148H221N41O47S
CAS Number:
Molecular Weight:
3358.65
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

[des-His1, Glu9]-Glucagon amide, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](Cc3ccccc3)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CO)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CO)NC(=O)[C@H](Cc5ccc(O)cc5)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](Cc6ccccc6)NC(=O)[C@@H](NC(=O)CNC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](N)CO)[C@@H](C)O)[C@@H](C)O)C(C)C)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(N)=O

InChI

1S/C148H221N41O47S/c1-70(2)54-95(131(221)171-94(49-53-237-11)130(220)179-102(61-111(154)202)140(230)188-117(74(8)194)120(155)210)174-135(225)101(60-81-64-162-86-29-19-18-28-84(81)86)178-128(218)92(43-47-110(153)201)172-144(234)116(72(5)6)187-138(228)99(56-77-24-14-12-15-25-77)177-136(226)103(62-114(206)207)180-127(217)91(42-46-109(152)200)165-121(211)73(7)164-124(214)88(31-22-51-160-147(156)157)167-125(215)89(32-23-52-161-148(158)159)169-142(232)106(68-192)184-137(227)104(63-115(208)209)181-132(222)96(55-71(3)4)173-133(223)97(58-79-33-37-82(197)38-34-79)175-126(216)87(30-20-21-50-149)168-141(231)105(67-191)183-134(224)98(59-80-35-39-83(198)40-36-80)176-129(219)93(44-48-113(204)205)170-143(233)107(69-193)185-146(236)119(76(10)196)189-139(229)100(57-78-26-16-13-17-27-78)182-145(235)118(75(9)195)186-112(203)65-163-123(213)90(41-45-108(151)199)166-122(212)85(150)66-190/h12-19,24-29,33-40,64,70-76,85,87-107,116-119,162,190-198H,20-23,30-32,41-63,65-69,149-150H2,1-11H3,(H2,151,199)(H2,152,200)(H2,153,201)(H2,154,202)(H2,155,210)(H,163,213)(H,164,214)(H,165,211)(H,166,212)(H,167,215)(H,168,231)(H,169,232)(H,170,233)(H,171,221)(H,172,234)(H,173,223)(H,174,225)(H,175,216)(H,176,219)(H,177,226)(H,178,218)(H,179,220)(H,180,217)(H,181,222)(H,182,235)(H,183,224)(H,184,227)(H,185,236)(H,186,203)(H,187,228)(H,188,230)(H,189,229)(H,204,205)(H,206,207)(H,208,209)(H4,156,157,160)(H4,158,159,161)/t73-,74+,75+,76+,85-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,116-,117-,118-,119-/m0/s1

InChI key

RVXSASLSNHDASC-OSWDIKPLSA-N

Amino Acid Sequence

Ser-Gln-Gly-Thr-Phe-Thr-Ser-Glu-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-NH2

Biochem/physiol Actions

The glucagon amide peptide [des-His1,Glu9]-glucagon amide is a glucagon receptor-adenyl cyclase activating antagonist that blocks glucagon-induced hyperglycemia and enhances glucose-stimulated pancreatic insulin release. Des-His1,Glu9]-glucagon amide is sometimes used in comparison with the glucagon agonists such as Lys17,18,Glu21-glucagon.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S ThanThan et al.
Domestic animal endocrinology, 42(3), 155-164 (2011-12-14)
Oxyntomodulin (OXM), glucagon, and glucagon-like peptide-1 (GLP-1), peptide hormones derived from the glucagon gene, play an important role in glucose homeostasis. The insulinotropic action of these three homologous peptides has been well documented in monogastric animals. However, information on the
Marita P Feldkaemper et al.
Visual neuroscience, 19(6), 755-766 (2003-04-12)
Eye growth and refraction are regulated by visual processing in the retina. Until now, the messengers released by the retina to induce these changes are largely unknown. Previously, it was found that glucagon amacrine cells respond to defocus in the
H Kofod et al.
International journal of peptide and protein research, 32(6), 436-440 (1988-12-01)
Glucagon and secretin and some of their hybrid analogs potentiate glucose-induced release of insulin from isolated mouse pancreatic islets. It was recently shown that the synthetic glucagon analog, desHis1[Glu9]glucagon amide, does not stimulate the formation of cyclic adenosine monophosphate in
C G Unson et al.
Peptides, 10(6), 1171-1177 (1989-11-01)
Hyperglycemia in diabetes mellitus is generally associated with elevated levels of glucagon in the blood. A glucagon analog, des-His1[Glu9]glucagon amide, has been designed and synthesized and found to be an antagonist of glucagon in several systems. It has been a
Xiaosong Ma et al.
Molecular endocrinology (Baltimore, Md.), 19(1), 198-212 (2004-10-02)
Glucagon, secreted by the pancreatic alpha-cells, stimulates insulin secretion from neighboring beta-cells by cAMP- and protein kinase A (PKA)-dependent mechanisms, but it is not known whether glucagon also modulates its own secretion. We have addressed this issue by combining recordings

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service