Skip to Content
Merck
All Photos(1)

Key Documents

G1127

Sigma-Aldrich

Gly-Gly hydrochloride

≥99%

Synonym(s):

(Glycylglycine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O3 · HCl
CAS Number:
Molecular Weight:
168.58
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

Gly-Gly hydrochloride,

Assay

≥99%

form

powder

technique(s)

electrophoresis: suitable

color

white

SMILES string

O.Cl.NCC(=O)NCC(O)=O

InChI

1S/C4H8N2O3.ClH.H2O/c5-1-3(7)6-2-4(8)9;;/h1-2,5H2,(H,6,7)(H,8,9);1H;1H2

InChI key

KHYIJCMSLMRPDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Gly

Application

Glycylglycine (Gly-Gly) dipeptide is used in the preparation of biodegradable polymers. Glycylglycine is used as a running buffer in capillary electrophoresis and capillary isotachophoresis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic Studies and Mechanism of Hydrogen Peroxide Catalytic Decomposition by Cu(II) Complexes with Polyelectrolytes Derived from L-Alanine and Glycylglycine.
Skounas S, Methenitis C, et al.
Bioinorganic Chemistry and Applications, DOI: 10-DOI: 10 (2010)
František Kvasnička et al.
Electrophoresis, 32(9), 1090-1093 (2011-04-02)
Phytic acid (PA) and lower inositolphosphates (InsP(n) ) is the main storage form of phosphorus in grains or seeds. The content of PA and InsP(n) in different varieties of barley was analyzed by capillary isotachophoresis and online-coupled capillary isotachophoresis with
Angelo Zinellu et al.
Journal of separation science, 28(16), 2193-2199 (2005-12-02)
A new CE method for ascorbic acid (AA) and uric acid (UA) detection in human plasma has been developed. Analytes were resolved in less than 4 min by employing sodium glycylglycine (Glygly) as electrolyte run buffer at pH 8.0. Using
Salvatore Sotgia et al.
Analytical and bioanalytical chemistry, 395(8), 2577-2582 (2009-10-07)
A rapid and simple short-end injection capillary zone electrophoresis method was developed for the quantification of plasma uric acid. The separation was performed in an uncoated fused-silica capillary (50 microm ID, 60 cm total length, 10.2 cm effective length) by
Meng Deng et al.
Advanced functional materials, 20(17), 2743-2957 (2011-07-27)
Synthetic biodegradable polymers serve as temporary substrates that accommodate cell infiltration and tissue in-growth in regenerative medicine. To allow tissue in-growth and nutrient transport, traditional three-dimensional (3D) scaffolds must be prefabricated with an interconnected porous structure. Here we demonstrated for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service