Skip to Content
Merck
All Photos(1)

Key Documents

G0540

Sigma-Aldrich

Monoclonal Anti-Granulotyte Colony Stimulating Factor antibody produced in rat

clone 67604.111, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-G-CSF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

67604.111, monoclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

indirect ELISA: 0.5-1.0 μg/mL
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Csf3(12985)

Specificity

The antibody will neutralize the biological activity of recombinant mouse G-CSF. By immunoblotting and ELISA, the antibody shows no cross-reactivity with recombinant human G-CSF.

Immunogen

recombinant mouse G-CSF expressed in E. coli.

Physical form

Lyophilized from 0.2 μm filtered solution of phosphate buffered saline

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service