Skip to Content
Merck
All Photos(2)

Key Documents

G0142

Sigma-Aldrich

Gly-Phe p-nitroanilide

cathepsin c substrate

Synonym(s):

Gly-Phe-pNA

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$1,264.00
250 MG
R$10,213.00

R$1,264.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
10 MG
R$1,264.00
250 MG
R$10,213.00

About This Item

Empirical Formula (Hill Notation):
C17H18N4O4
CAS Number:
Molecular Weight:
342.35
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

R$1,264.00


Estimated to ship onMay 29, 2025


Assay

≥98% (TLC)

form

powder

solubility

DMF: 50 mg/mL, clear, yellow

storage temp.

−20°C

SMILES string

NCC(=O)NC(Cc1ccccc1)C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C17H18N4O4/c18-11-16(22)20-15(10-12-4-2-1-3-5-12)17(23)19-13-6-8-14(9-7-13)21(24)25/h1-9,15H,10-11,18H2,(H,19,23)(H,20,22)

InChI key

KNDZCZRECXTRHP-UHFFFAOYSA-N

Substrates

Chromogenic substrate for cathepsin C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple estimation of cathepsin C using a new chromogenic substrate.
R J PLANTA et al.
Analytical biochemistry, 5, 360-362 (1963-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service