Skip to Content
Merck
All Photos(2)

Key Documents

F9807

Sigma-Aldrich

FSL-GB3

Synonym(s):

FSL-GB3(GALa4GALb4GLCb)-SA1-L1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C68H122N2NaO26P
Molecular Weight:
1437.66
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥95% (TLC)

Quality Level

form

solid

storage temp.

−20°C

Application

All FSL constructs disperse in biocompatible media and spontaneously and stably incorporate into cell membranes. Cells modified with KODE constructs are known as kodecytes and usually maintain their normal vitality and functionality.

FSL-GB3 has been specifically designed to create live cells (kodecytes) expressing GB3 antigens but can also be used to modify other hydrophobic surfaces including fixed cells, virions, and solid phase surfaces. This product has been used in vitro to inhibit HIV virions and bind to veratoxin.
All FSL constructs disperse in biocompatible media and spontaneously and stably incorporate into cell membranes. FSL-GB3 has been specifically designed to create live cells (kodecytes) expressing GB3 antigens but can also be used to modify other hydrophobic surfaces including fixed cells, virions, and solid phase surfaces.

Other Notes

FSL-GB3 is a KODE technology construct designed to label hydrophobic surfaces including living cells with blood group GB3 trisaccharide (also known as Pk). All KODE FSL constructs consist of three essential designable features: a functional component (F), a spacer (S), and a diacyl lipid (L). FSL-GB3 is comprised of the blood group GB3 (Pk) trisaccharide Galα4Galβ4Glcβ representing F, conjugated via an O(CH2)3NH spacer (SA1) to an activated adipate derivative of dioleoyl­phos­pha­tidyl­ethanolamine (L1).

Legal Information

KODE Constructs are supplied by KODE Biotech Materials Limited under license from KODE Biotech Limited.
KODE is a trademark of KODE Biotech Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service