Skip to Content
Merck
All Photos(1)

Key Documents

F9052

Sigma-Aldrich

Anti-FRS2 (SNT-1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FRS1A, Anti-FRS2A, Anti-FRS2alpha, Anti-SNT, Anti-SNT-1, Anti-SNT1

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 ML
R$4,982.00

R$4,982.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

Select a Size

Change View
0.2 ML
R$4,982.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

R$4,982.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 92-95 kDa (doublet)

species reactivity

rat, mouse, human

technique(s)

immunoprecipitation (IP): 10-20 μg using RIPA lysate of mouse NIH/3T3 cells
microarray: suitable
western blot: 1:1,000 using whole cell extracts of rat PC-12 cells or human HepG2 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FRS2(10818)
mouse ... Frs2(327826)
rat ... Frs2(314850)

General description

Fibroblast growth factor receptor substrate 2 (FRS2), also known as suc-1 associated neurotrophic factor target protein (SNT) exist in two structurally similar forms namely, FRS2a (SNT-1) and FRS-2b (SNT-2). FRS2 is characterized with a consensus myristylation sequence, involved in its recruitment to the cell membrane 4 and a putative phosphotyrosine binding (PTB) domain in its amino-terminus.

Immunogen

synthetic peptide corresponding to human FRS2 sequence, (amino acids 487-505). The corresponding sequence is identical in Xenopus and differs from the respective human and mouse FRS2β sequences by 2 and 3 amino acids, respectively.

Biochem/physiol Actions

Fibroblast growth factors (FGF) receptor substrate (FRS) functions as a lipid-anchored docking protein that is tyrosine phosphorylated and recruited to FGFR upon FGF stimulation. fibroblast growth factor receptor substrate 2 (FRS2) plays an important role in linking fibroblast growth factor (FGF) and nerve growth factor (NGF) with the Ras/ mitogen-activated protein kinases (MAPKs) signaling pathway, thus relaying information from the cell surface to the nucleus. FRS2 proteins are tyrosine phosphorylated in response to activation of the RET receptor, a tyrosine kinase that functions as the signal transducing receptor for the glial cell line-derived neurotrophic factor (GDNF).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Docking protein FRS2 links the protein tyrosine kinase RET and its oncogenic forms with the mitogen-activated protein kinase signaling cascade
Melillo RM, et al.
Molecular and Cellular Biology, 21(13), 4177-4187 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service