Skip to Content
Merck
All Photos(1)

Key Documents

F8507

Sigma-Aldrich

(±)-Fenfluramine hydrochloride

Synonym(s):

(±)-N-Ethyl-α-methyl-m-[trifluoromethyl]phenethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16F3N · HCl
CAS Number:
Molecular Weight:
267.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

drug control

USDEA Schedule IV

application(s)

forensics and toxicology

SMILES string

Cl.CCNC(C)Cc1cccc(c1)C(F)(F)F

InChI

1S/C12H16F3N.ClH/c1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15;/h4-6,8-9,16H,3,7H2,1-2H3;1H

InChI key

ZXKXJHAOUFHNAS-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Neurotoxic on prolonged administration or at high dosage; releases serotonin from axon terminals by a nonexocytotic mechanism; anorectic

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y X Wang et al.
The Journal of pharmacology and experimental therapeutics, 291(3), 1008-1016 (1999-11-24)
Fenfluramine is an indirect agonist of 5-hydroxytryptamine (5-HT) receptors that acts by evoking 5-HT release and blocking 5-HT reuptake in neuronal cells. The current study compared the antinociceptive properties of fenfluramine with those of the tricyclic antidepressants amitriptyline and desipramine
Justin N Siemian et al.
British journal of pharmacology, 175(9), 1519-1534 (2018-02-17)
Although the antinociceptive efficacies of imidazoline I2 receptor agonists have been established, the exact post-receptor mechanisms remain unknown. This study tested the hypothesis that monoaminergic transmission is critical for I2 receptor agonist-induced antinociception. von Frey filaments were used to assess
J D Roth et al.
European journal of pharmacology, 373(2-3), 127-134 (1999-07-22)
Fenfluramine + phentermine was a widely used combination for weight loss. Fenfluramine and phentermine are believed to act via serotonin and catecholamines, respectively. To what extent these drugs interact has not been well-established. We compared the anorectic efficacy of a
Eileen K Sawyer et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 37(8), 1816-1824 (2012-03-22)
Acute SSRI (selective serotonin reuptake inhibitor) treatment has been shown to attenuate the abuse-related effects of cocaine; however, SSRIs have had limited success in clinical trials for cocaine abuse, possibly due to neurobiological changes that occur during chronic administration. In
Geneviève Derumeaux et al.
PloS one, 7(6), e38273-e38273 (2012-06-23)
REGULATE trial was designed to compare the efficacy and safety of benfluorex versus pioglitazone in type 2 diabetes mellitus (DM) patients. Double-blind, parallel-group, international, randomised, non-inferiority trial. More than half of the 196 participating centres were primary care centres. Patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service