Skip to Content
Merck
All Photos(1)

Key Documents

F7930

Sigma-Aldrich

FGFR3 (397-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Eph receptor tyrosine kinase, growth hormone receptor, ACH, CD333, CEK2, HSFGFR3EX, JTK4

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$4,769.00

R$4,769.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
R$4,769.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

R$4,769.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

95-129 nmol/min·mg

mol wt

~73 kDa

packaging

pkg of 10 μg

technique(s)

cell based assay: suitable

solubility

water: soluble

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FGFR3(2261)

General description

Research area: Cancer


FGFR3 (fibroblast growth factor receptor 3) is a receptor tyrosine kinase (RTK) and belongs to the FGFR subfamily. This subfamily includes four members namely, FGFR1, FGFR2, FGFR3, and FGFR4. It contains a ligand-binding region in its exoplasmic domain, a single transmembrane region, and a tyrosine kinase domain in its intracellular domain.[1]FGFR3 has a strong affinity for canonical FGF ligands.[2]

Biochem/physiol Actions

FGFR3 (fibroblast growth factor receptor 3) plays an essential role in the signaling pathways during embryogenesis and angiogenesis. [3] This gene is either mutated or up-regulated in multiple myeloma, cervical carcinoma [3] and nonmuscle-invasive urothelial carcinoma (UC) of the bladder.[4]Apart from this, certain mutations in FGFR3 are also known to be linked with skeletal disorders such as various forms of dwarfism. [3] FGFR3 also plays hepatoprotective function during liver damage. [5]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corine G M L'Hôte et al.
Experimental cell research, 304(2), 417-431 (2005-03-08)
FGFR3 is a receptor tyrosine kinase (RTK) of the FGF receptor family, known to have a negative regulatory effect on long bone growth. Fgfr3 knockout mice display longer bones and, accordingly, most germline-activating mutations in man are associated with dwarfism.
Hiroko Tamagaki et al.
Biochemistry, 53(30), 5000-5007 (2014-07-11)
Activation of the protein tyrosine kinase receptors requires the coupling of ligand binding to a change in both the proximity and orientation of the single transmembrane (TM) helices of receptor monomers to allow transphosphorylation of the receptor kinase domain. We
FGFR3 biology and skeletal disease
Jyoti Narayana and William A Horton
Connective tissue research, 56(6), 427-433 (2015)
Lin Chen et al.
Frontiers in bioscience : a journal and virtual library, 10, 1961-1976 (2005-03-17)
Fibroblast growth factor receptors (FGFRs) exist as a gene family of 4 membrane bound receptor tyrosine kinases (FGFR1-4) that mediate signals of at least 22 fibroblast growth factors (FGF1-22). FGFs/FGFRs play important roles in multiple biological processes, including mesoderm induction
Elizabeth A Guancial et al.
Cancer medicine, 3(4), 835-844 (2014-05-23)
While fibroblast growth factor receptor 3 (FGFR3) is frequently mutated or overexpressed in nonmuscle-invasive urothelial carcinoma (UC), the prevalence of FGFR3 protein expression and mutation remains unknown in muscle-invasive disease. FGFR3 protein and mRNA expression, mutational status, and copy number

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service