F4757
D-Fructose 1,6-bisphosphate sodium salt hydrate
≥70%
Synonym(s):
D(+)Fructofuranose 1,6-diphosphate, Harden-Young ester, Hexose diphosphate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H14O12P2 · xNa+ · yH2O
CAS Number:
Molecular Weight:
340.12 (anhydrous free acid basis)
EC Number:
UNSPSC Code:
12352201
PubChem Substance ID:
Recommended Products
Assay
≥70%
storage temp.
−20°C
SMILES string
O.[Na+].[Na+].[Na+].[Na+].O[C@H]1[C@H](O)[C@@](O)(COP([O-])([O-])=O)O[C@@H]1COP([O-])([O-])=O
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yasmean Kalam et al.
Clinical toxicology (Philadelphia, Pa.), 50(7), 546-554 (2012-08-09)
Fructose-1,6-diphosphate (FDP) is a metabolite in the glycolytic pathway created from glucose. Exogenously administered FDP increases the yield of ATP from anaerobic glycolysis. FDP reduces ischaemic tissue area in experimentally-induced cerebral and myocardial infarction and improves haemodynamics post-cardiac bypass. We
Ming Xu et al.
The Journal of pharmacy and pharmacology, 64(2), 244-251 (2012-01-10)
Phosphodiesterase type 5 inhibitors are potent in relieving erectile dysfunction (ED), however, they are less satisfactory in diabetic patients, probably due to the pro-inflammatory biomarkers caused by diabetes. Therefore, it was interesting to compare the effects of sildenafil with strontium
Laura Novellasdemunt et al.
The Journal of biological chemistry, 288(15), 10640-10651 (2013-03-05)
Reciprocal regulation of metabolism and signaling allows cells to modulate their activity in accordance with their metabolic resources. Thus, amino acids could activate signal transduction pathways that control cell metabolism. To test this hypothesis, we analyzed the effect of amino
Andreas Neuner et al.
Journal of biotechnology, 163(2), 217-224 (2012-08-18)
Corynebacterium glutamicum, the best established industrial producer organism for lysine was genetically modified to allow the production of lysine on grass and corn silages. The resulting strain C. glutamicum lysC(fbr)dld(Psod)pyc(Psod)malE(Psod)fbp(Psod)gapX(Psod) was based on earlier work (Neuner and Heinzle, 2011). That
Thomas J Wheeler et al.
Molecular and cellular biochemistry, 366(1-2), 31-39 (2012-03-20)
Previously, we reported that fructose-1,6-bisphosphate (FBP) was taken up by rat cardiac myocytes by two processes: a component that was saturable at micromolar levels and a nonsaturable component that dominated at millimolar levels. Here, we continued to characterize the saturable
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service