Skip to Content
Merck
All Photos(1)

Documents

F3526

Supelco

Fluorescent Molecular Weight Marker

mol wt 20,000-200,000 Da

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

lyophilized powder

mol wt

20,000-200,000 Da

color

blue

storage temp.

−20°C

Other Notes

This product contains the following proteins:
  • Trypsin Inhibitor, Soybean: 20,100 Da
  • Carbonic Anhydrase, Bovine Erythrocyte: 29,000 Da
  • Alcohol Dehydrogenase, Horse Liver: 39,800 Da
  • Albumin, Bovine Serum: 66,000 Da
  • β-Galactosidase, E. coli: 116,000 Da
  • Myosin, Rabbit Muscle: 205,000 Da

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo L V Silveira et al.
Clinical and vaccine immunology : CVI, 15(8), 1292-1300 (2008-06-27)
Earlier studies have demonstrated in A/Sn mice highly susceptible to Chagas' disease protective immunity against lethal Trypanosoma cruzi infection elicited by vaccination with an open reading frame (ORF) expressed by amastigotes. In our experiments, we used this mouse model to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service