Skip to Content
Merck

Skip To

F2001

5-Fluoroindole-3-carboxaldehyde

≥99%, crystalline

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C9H6FNO
CAS Number:
Molecular Weight:
163.15
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352108
MDL number:
Assay:
≥99%
Form:
crystalline


assay

≥99%

form

crystalline

color

colorless to yellow

storage temp.

−20°C

SMILES string

Fc1ccc2[nH]cc(C=O)c2c1

InChI

1S/C9H6FNO/c10-7-1-2-9-8(3-7)6(5-12)4-11-9/h1-5,11H

InChI key

YUAJKGBLPVLADK-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters