Skip to Content
Merck
All Photos(1)

Key Documents

F1928

Sigma-Aldrich

Farnesyl triphosphate ammonium salt solution

≥95% (TLC), methanol:ammonia solution

Synonym(s):

3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl triphosphate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29O10P3 · 4NH3
Molecular Weight:
530.43
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥95% (TLC)

form

methanol:ammonia solution

packaging

vial of 200 μg

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[NH4+].[NH4+].[NH4+].[NH4+].C\C(C)=C\CC\C(C)=C\CC\C(C)=C\COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

InChI

1S/C15H29O10P3.4H3N/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-23-27(19,20)25-28(21,22)24-26(16,17)18;;;;/h7,9,11H,5-6,8,10,12H2,1-4H3,(H,19,20)(H,21,22)(H2,16,17,18);4*1H3/b14-9+,15-11+;;;;

InChI key

PKNKGXPLZFBJFK-OBMCCXAOSA-N

Other Notes

Possibly involved in isoprenoid metabolism

Physical form

Solution in methanol:10 mM ammonium hydroxide (7:3)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service