Skip to Content
Merck
All Photos(1)

Key Documents

F0502

Sigma-Aldrich

D-Fructose 1,6-bisphosphate dicalcium salt

practical grade, ~70%

Synonym(s):

D(+)Fructofuranose 1,6-diphosphate, Harden-Young ester, Hexose diphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10Ca2O12P2
CAS Number:
Molecular Weight:
416.24
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

practical grade

Assay

~70%

form

solid

SMILES string

[Ca].OC(COP(O)(O)=O)C(O)C(O)C(=O)COP(O)(O)=O

InChI

1S/C6H14O12P2.Ca.2H/c7-3(1-17-19(11,12)13)5(9)6(10)4(8)2-18-20(14,15)16;;;/h3,5-7,9-10H,1-2H2,(H2,11,12,13)(H2,14,15,16);;;

InChI key

KJZXQSWXBTUHIN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasmean Kalam et al.
Clinical toxicology (Philadelphia, Pa.), 50(7), 546-554 (2012-08-09)
Fructose-1,6-diphosphate (FDP) is a metabolite in the glycolytic pathway created from glucose. Exogenously administered FDP increases the yield of ATP from anaerobic glycolysis. FDP reduces ischaemic tissue area in experimentally-induced cerebral and myocardial infarction and improves haemodynamics post-cardiac bypass. We
Ming Xu et al.
The Journal of pharmacy and pharmacology, 64(2), 244-251 (2012-01-10)
Phosphodiesterase type 5 inhibitors are potent in relieving erectile dysfunction (ED), however, they are less satisfactory in diabetic patients, probably due to the pro-inflammatory biomarkers caused by diabetes. Therefore, it was interesting to compare the effects of sildenafil with strontium
Gauri S Joshi et al.
Journal of bacteriology, 194(6), 1350-1360 (2012-01-17)
The cbb(I) region of Rhodopseudomonas palustris (Rp. palustris) contains the cbbLS genes encoding form I ribulose-1,5-bisphosphate (RuBP) carboxylase oxygenase (RubisCO) along with a divergently transcribed regulator gene, cbbR. Juxtaposed between cbbR and cbbLS are the cbbRRS genes, encoding an unusual
Guilherme Vargas Bochi et al.
Inflammation, 35(6), 1786-1792 (2012-07-11)
The accumulation of advanced oxidation protein products (AOPP) has been linked to several pathological conditions. Previous studies have identified AOPP as a novel biomarker of oxidative damage to proteins and a novel class of mediator of inflammation. The aim of
Roberto Christ Vianna Santos et al.
Inflammation, 35(3), 1198-1203 (2012-02-14)
It has been previously showed that fructose-1,6-bisphosphate (FBP) has anti-inflammatory and immunomodulatory effects on several experimental inflammation models. However, the effects and mechanism of FBP on Zymosan-induced acute lung injury (ALI) in mice had not been tested. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service