Skip to Content
Merck
All Photos(1)

Key Documents

EPM1194-9907

Platform for Eppendorf® New Brunswick Innova® 2100/40/40R/42/42R & Excella® E24/E24R Shakers

for 1 L Erlenmeyer flasks, dedicated, phenolic resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103816
NACRES:
NB.73
Pricing and availability is not currently available.

feature

9 x 1 L Erlenmeyer flasks
dedicated
for 1 L Erlenmeyer flasks
phenolic resin

manufacturer/tradename

Eppendorf® M1194-9907 (Global)

L × W

45.7 cm × 45.7 cm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Platforms designed for the Eppendorf New Brunswick Innova 2100/40/40R/42/42R & Excella E24/E24R shakers to ensure a safe and perfect fit.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service