EP6334000042
Eppendorf® Mastercycler® nexus
Gradient Eco, satellite unit, aluminum block, AC/DC input 230V (GB plug)
About This Item
feature
to hold 96 x 0.1mL PCR tubes OR 96 x 0.2mL PCR tubes OR 71 x 0.5mL PCR tubes OR 1 PCR plate (96 wells) OR 38x0.5mL Eppendorf PCR tubes
CSA and CE compliant
Gradient Eco
aluminum block
gradient block
satellite unit
unit can only be controlled
manufacturer/tradename
Eppendorf® 6334000042 (GB)
parameter
1-20 °C (gradient range)
37-110 °C temp. range (Lid)
4-99 °C temp. range (control range)
4-99 °C temp. range
±0.2 °C temperature
W × D × H
25 cm × 41.2 cm × 32.1 cm
weight
10.4 kg
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Small footprint
- Universal block for ultimate flexibility
- Available with gradient
- E-mail notification
- flexlid® concept allows use of all types of consumables with automatic height adjustment of the lid
Legal Information
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service