Key Documents
EP5427739009
Eppendorf® Fixed-angle rotor
F-35-6-30, incl. rotor lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Pricing and availability is not currently available.
feature
to hold 12 x Vacutainer® tubes
to hold 6 x 15/50 mL tubes
F-35-6-30, incl. rotor lid
packaging
pack of 1 ea
pkg of 1 ea
manufacturer/tradename
Eppendorf® 5427739009
parameter
7,830 max. RCF (7,745 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Fixed angle rotors for use with Eppendorf centrifuge models.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)