Key Documents
EP5392000013
Eppendorf(R) HeatSealer
S200
About This Item
UNSPSC Code:
41102400
Pricing and availability is not currently available.
AC/DC input
100 - 230 V, 50/60 Hz, US 2-pin plug
feature
CE compliant
CSA approved
adjustable Sealing, temperature
seals 96 and 384 well plates
manufacturer/tradename
Eppendorf® 5392000013 (US)
General description
The Eppendorf HeatSealer Family offers you reliable and effective Sealing, of a variety of plates. You can choose from a more compact (S100) and a more flexible (S200) instrument according to your Sealing, needs.
Features and Benefits
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)