EP5383000019
Eppendorf® ThermoStat C
basic device without thermoblock, AC/DC input 220 - 240 V, 50/60 Hz (EU)
About This Item
Recommended Products
AC/DC input
, EuroPlug
feature
CE compliant
USB Interface
manufacturer/tradename
Eppendorf® 5383000019
temp measuring range
accuracy: ±0.5 °C (at 20-45 °C)
parameter
-10-110 °C temp. range
W × D × H
20.6 cm × 30.4 cm × 13.6 cm
weight
4.3 kg
Looking for similar products? Visit Product Comparison Guide
General description
Eppendorf ThermoTop condens.protect
Now you can control the temperature of your samples with ease: The ThermoTop, with unique condens.protect technology, reliably prevents condensation formation on the tube lid and wall.
- Efficient and reliable condensation prevention on tube lid and wall
- Guarantees constant reaction conditions for optimal enzymatic performance
- Improves handling times, tubes don‘t have to be spun down at the end of the incubation
- Optionally available for Eppendorf SmartBlocks 0.5/1.5 and 2.0 mL as well as Eppendorf SmartBlocks plates and PCR 96 & PCR 384 – labeled with the condens.protect symbol
A wide varity of Eppendorf SmartBlocks are available for the Eppendorf ThermoStat C. These thermobocks offer the utmost flexibility for use of all common vessel and plate formats.
- Fast and simple block exchange due to Eppendorf QuickRelease
- Optimized block design guarantees maximum temperature transfer to the sample
- Individually sensor?controlled Eppendorf SmartBlocks allow for block-specific calibration
Features and Benefits
- Heating and active cooling in all common vessel and plate formats from 5 μL to 50 mL for high flexibility
- Broad range of SmartBlocks enables high vessel flexibility
- Excellent temperature accuracy for safe sample handling
- Very fast cooling rates of up to 5 deg. C/min, reducing your waiting time
- Low temperature (e.g. 4 deg. C or 0 deg. C) even at higher ambient temperatures for safe sample incubation
- Temperature keys (4 deg. C, 16 deg. C, 37 deg. C, 56 deg. C, and 95 deg. C) provide quick access to commonly used experimental temperatures
- Eppendorf ThermoTop (condens.protect) compatibility prevents condensation and improves temperature homogeneity within sample tubes (up to 2.0 mL)
- Comprehensive range of program slots (up to 15) for flexible usage of device
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service